missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution
Buffer cell culture with PBS in solution.
Brand: Cytiva SH30378.03
This item is not returnable.
View return policy
Description
- Format: Liquid

Specifications
Manufactured under cGMP and ISO 9001 certified facilities | |
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) Without Calcium, Magnesium, Phenol Red | |
Liquid | |
Sterile | |
None | |
≤0.025EU/mL | |
276±10mOsm/kg H2O | |
1 L |
15° to 30°C | |
10 X | |
Dulbecco's Phosphate Buffered Saline | |
0.1 μm Sterile-filtered | |
No Calcium, No Magnesium, No Phenol Red | |
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
7 to 7.6 | |
RUO |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN AQUATIC-Contains 3% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction