missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)
Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.
Brand: Cytiva SH30084.03IR30-45
1206.50 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Filtered through 3 sequential 0.1 μm pore size filters
Specifications
Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Fetal Bovine Serum | |
Sterile | |
Characterized | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
HyClone™ |
AQIS | |
Australia | |
500 mL | |
Sequential Triple-filtered, 100 nm | |
Irradiated (30 to 45 doses) | |
No | |
USDA Inspected |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction