missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)

Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.

Brand:  Cytiva SH30084.03IH30-45

 View more versions of this product

Product Code. 17276896

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Filtered through 3 sequential 0.1 μm pore size filters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Liquid
500 mL
Sequential Triple-filtered, 100 nm, True Pool Technology
Heat Inactivated, Irradiated (30 to 45 doses)
Yes
USDA Inspected
AQIS
Australia
Fetal Bovine Serum
Sterile
Characterized
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
HyClone™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.