missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

147.90 € - 1215.00 €

Specifications

Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Product Type Fetal Bovine Serum
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
View More Specs

Products 8
Product Code Brand Quantity Processing Price Quantity & Availability  
Product Code Brand Quantity Processing Price Quantity & Availability  
11521831
View Documents
Cytiva
SH30071.02
100 mL None
152.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
17296886
View Documents
Cytiva
SH30071.02IH25-40
100 mL Heat Inactivated, Irradiated (25 to 40 doses) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17206896
View Documents
Cytiva
SH30071.02IR25-40
100 mL Irradiated (25 to 40 doses) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11531831
View Documents
Cytiva
SH30071.03
500 mL None
1215.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
12379802
View Documents
Cytiva
SH30071.03HI
500 mL Heat Inactivated
667.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11306060
View Documents
Cytiva
SH30071.03IR
500 mL Irradiated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16281082
View Documents
Cytiva
SH30071.03IR25-40
500 mL Irradiated (25 to 40 doses)
657.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16291082
View Documents
Cytiva
SH30071.03IR30-45
500 mL Irradiated (30 to 45 doses)
657.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100 nm pore-size rated filters
Specifications

Specifications

Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Fetal Bovine Serum
Sequential Triple-filtered, 100 nm
≤ 25 EU/mL
HyClone™
USDA Inspected
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Sterile
Characterized
≤ 25 mg/dL
No
Videos
SDS
Documents

Documents

Product Certifications