missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated
Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.
Brand: Cytiva SH30068.03HI
692.55 EUR valid until 2025-12-31
Use promo code "24268" to get your promotional price.
This item is not returnable.
View return policy
Description
- Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
- This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
- Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
- Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications
1, 25 Dihydroxy Vitamin D, 17 Beta Estradiol, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcitonin, Calcium, Carotene, Chloride, Corticosterone, Cortisol, Creatinine, Estrone, Folate, FSH, Glucose, IgG, Inorganic Phosphorus, Insulin, Lactate Dehydrogenase, LH, Niacin, Potassium, Progesterone, Retinol, SGOT, SGPT, Sodium, T3, T4, Testosterone, Tocopherol, Total Bilirubin, Total Protein, TSH, Vitamin C | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
USDA Inspected |
EMEA Certified | |
Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, Heat Inactivated | |
Charcoal/Dextran filtered | |
Sterile | |
Specialty filtered | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
No |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-2.1 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 2.1% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction