missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Processing:
Gamma Irr, 25 to 40 doses
Gamma Irradiated
HI, Irr - 25 to 40 doses
Heat Inactivated
None
Unit Size:
100mL
500mL
50mL
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
Specifications
| Additional Assays | 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1 |
| Certifications/Compliance | EMEA Certified |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Country of Origin | United States |
| Specialty Type | Dialyzed |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Grade | Specialty filtered |
| Processing | Gamma Irr, 25 to 40 doses |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction