missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ I Serum (U.S.)

Product Code. 10275022
Click to view available options
Quantity:
100 mL
500 mL
Heat Inactivated:
No
Yes
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Produktkode 10275022

missing translation for 'mfr': Cytiva SH30080.02

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • Demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • TRUSTED_SUSTAINABILITY

    Tekniske data

    Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
    Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
    Description FetalClone I Serum (U.S.)
    Country of Origin United States
    Product Type FBS Replacement Serum
    Quantity 100 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 10 EU/mL
    Heat Inactivated No
    Hemoglobin Concentration ≤ 20 mg/dL
    IgG levels Typically less than 2μg/mL
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Vis mere Vis mindre
    Korrektion af produktindhold

    Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

    Produkttitel

    Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

    Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.