missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Buffer cell culture growth with 1X balanced salt solutions
Brand: Cytiva SH30031.03
This item is not returnable.
View return policy
Description
- Used for washing and treating tissues and cells
Specifications
cGMP and ISO 9001:2000 Guidelines | |
1 X | |
Hanks' Balanced Salt Solution | |
Phenol Red | |
=1.0EU/mL | |
1000 mL |
15° to 30°C | |
Liquid | |
0.1 μm Sterile-filtered | |
No Calcium, No Magnesium | |
7.0 to 7.4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions > Liquid; Without Calcium, Magnesium/With Phenol Red; 1000mL
Spot an opportunity for improvement?Share a Content Correction