Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Buffer cell culture growth with 1X balanced salt solutions
Brand:  Cytiva SH30031.02
16.96 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 10532003
  • 24.40 € / 500mL
Explore more special offers
Description
Description
  • Used for washing and treating tissues and cells
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cGMP and ISO 9001:2000 Guidelines
1 X
Hanks' Balanced Salt Solution
Phenol Red
=1.0EU/mL
500 mL
15° to 30°C
Liquid
0.1 μm Sterile-filtered
No Calcium, No Magnesium
7.0 to 7.4