Learn More
Cytiva HyClone™ Hank's Balanced Salt Solution Powder
Available in lot sizes that facilitate scale-up and manufacturing.
Brand:  Cytiva SH30016.02
26.60 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 12329722
  • 36.85 € / Pack of 2
Explore more special offers
Description
Description
HyClone™ Hanks' Balanced Salt Solution Powder is available in lot sizes that facilitate scale-up and manufacturing.
  • Cell culture application
  • All formulations incorporate a rigorous specification and approval process
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    cGMP and ISO 9001:2000 Guidelines
    Phenol Red
    2 x 5 L
    Without Calcium, without Magnesium, with Phenol Red (powder)
    No Calcium, No Magnesium