Learn More
Cytiva HyClone™ Heat Inactivated Characterized Fetal Bovine Serum (New Zealand)
Carefully collected, processed, and filtered. Cytiva HyClone™ Heat Inactivated Characterized Fetal Bovine Serum (New Zealand) is filtered through three sequential 0.1μm pore size-rated filters. Before dispensing, each lot is pooled to ensure uniformity and consistency between bottles.
Brand: Cytiva SH30406.02HI
1026.00 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
New Zealand’s islands are protected from many outside influences, both geographically and biologically. One benefit of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
- Complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology, which minimizes bottle-to-bottle variability
Specifications
Progesterone, Chloride, Insulin, Inorganic Phosphorus, PTH, Glucose, Cholesterol, Copper, HDL, Magnesium, LDL, Selenium, Triglycerides, IgG, Gamma Globulin, pH, Phospholipids, Osmolality, Alkaline Phosphatase, Iron, Lactate Dehydrogenase, TIBC, SGPT, Percent Saturation, SGOT, B1, B2, B6, Niacin, Biotin, Retinol, Carotene, Tocopherol, B12, Folate, Vitamin C, 1, 25 Dihydroxy Vitamin D, Total Protein, Albumin, Blood Urea Nitrogen | |
Solution | |
500 mL | |
Sequential Triple-filtered, 100 nm | |
≤20 EU/mL | |
Yes |
Store at <–10°C; once thawed, store at 2–8°C. | |
Fetal Bovine Serum | |
Sterile | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Cell Culture | |
≤25mg/dL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.