Learn More
Cytiva HyClone™ HyCell™ CHO Media, Powder
A versatile, animal-derived component-free (ADCF) cell culture media
Brand:  Cytiva SH30948.01
Product Code. 17612085
  • 304.00 € / 5 L
Explore more special offers
Description
Description
  • Developed for Chinese Hamster Ovary (CHO) Cell Cultures in bioprocessing applications
  • Regulatory-friendly, chemically defined, and animal-derived component-free media
  • Specifically formulated through the HyClone™ Metabolic Pathway Design process
  • Provides excellent performance, supports high peak cell density, and enhances product yield and quality
  • Media can be used for batch, fed-batch, or perfusion applications in multiple cell culture platforms
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Complete Powder without Poloxamer 188, L-Glutamine, Hypoxanthine, and Thymidine
CHO Medium
0.1 μm Sterile-filtered
5 L
Store at 2°C to 8°C, Away from Light
Powder
Sterile
HyCell™