missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum, New Zealand Origin
Enhances cell culture growth, contains three to four times as much available iron and transferrin as FBS or equine serum
Brand: Cytiva SH30626.02HI
Code : 84
Additional Details : Weight : 0.80000kg
Description
- Tested for endotoxin and hemoglobin levels
- Growth promotion studies have demonstrated various product applications
- Often outperforms FBS in many applications
- Sourced from animals that are typically less than 10 days old
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGTP), Glutamic Oxaloacetic Transaminase (SGOT), Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorous, Glucose, pH, Osmolality, Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, and IgG | |
New Zealand | |
Newborn Calf Bovine Serum | |
Sterile | |
Iron Supplemented, Heat Inactivated | |
Yes |
<10 days | |
Frozen | |
500 mL | |
Sequential-filtered, 100 nm | |
Fluorescent Antibody, Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine Respiratory Syncytial Virus, Bovine Viral Diarrhea Virus, Rabies, Reovirus, Cytopathogenic Agents (e.g., IBR), and Hemadsorbing agents (e.g., PI3) | |
HyClone™ |