missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iscove's Modified Dulbecco's Medium (IMDM)

Support cell culture growth and biomanufacturing processes with HyClone™ Iscove's Modified Dulbecco's Medium (IMDM), manufactured using ISO 9001:2000-certified processes.

Brand:  Cytiva SH30259.01

 View more versions of this product

Product Code. 10275212

  • 33.45 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
  • All products are hydrated using purified process water and undergo sterile filtration
  • Manufactured using ISO 9001 certified processes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 9001:2000
Without L-Glutamine
IMDM with HEPES; without L-glutamine
0.1 μm Sterile-filtered
No Glutamine
2° to 8°C, away from light
Liquid
Sterile-filtered
HEPES
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.