missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iscove's Modified Dulbecco's Medium (IMDM)

Support cell culture growth and biomanufacturing processes with HyClone™ Iscove's Modified Dulbecco's Medium (IMDM), manufactured using ISO 9001:2000-certified processes.

Brand:  Cytiva SH30228.02

27.62 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10386852

  • 33.40 € / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
  • All products are hydrated using purified process water and undergo sterile filtration
  • Manufactured using ISO 9001 certified processes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 9001:2000
With L-Glutamine, with HEPES, without Alpha-Thioglycerol
IMDM modified medium with L-glutamine, HEPES
0.1 μm Sterile-filtered
No α-Thioglycerol
2° to 8°C, away from light
Liquid
Sterile-filtered
Glutamine, HEPES
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers