Learn More
Cytiva HyClone™ Minimal Essential Medium with Earle's (MEM/EBSS), Powder
Brand:  Cytiva SH30050.03
 View more versions of this product
Product Code. 11551821
  • 38.60 € / 10 L
Explore more special offers
Description
Description
Support cell culture growth and biomanufacturing processes with powdered HyClone™ Minimal Essential Medium with Earle's (MEM/EBSS), manufactured using ISO 9001:2000-certified processes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cGMP and ISO 9001:2000
MEM/EBSS with L-Glutamine, Nonessential Amino Acids
MEM/EBSS with L-glutamine, nonessential amino acids (NEAA)
Glutamine, Ribonucleosides, Deoxyribonucleosides
10 L
2° to 8°C, away from light and moisture
Powder
Non-sterile
No Sodium Bicarbonate