missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Porcine Serum (New Zealand)

Product Code. 16695785
Click to view available options
Quantity:
1 L
1 L (QC Samples)
Processing:
Irradiated
Non-irradiated
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 16695785

Brand: Cytiva SH30908.04NI

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used to enhance cell culture performance, cost-efficient alternatives to Fetal Bovine Serum (FBS)

Cytiva's porcine serum is sourced from New Zealand, filtered in the U.S. through multiple 200 nm pore size-rated filters and is gamma irradiated at 25 to 40 kGy after filtration.
TRUSTED_SUSTAINABILITY

Specifications

Country of Origin New Zealand
Product Type Porcine Serum
Quantity 1 L
Sterilization Method Sequential Triple-filtered, 200 nm
Processing Non-irradiated
Endotoxin Concentration ≤ 100 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 50 mg/dL
Product Line HyClone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.