missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated

Augment cell culture growth with this cost-effective alternative to Fetal Bovine Serum (FBS)

Brand:  Cytiva SH30118.04HI

 View more versions of this product

Product Code. 12654207

  • 255.00 € / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Produced using careful collection and processing procedures, including venipuncture, to provide the highest quality product available
  • Ensures excellent culture results without fear of contamination
  • HyClone™ maintains complete control of the entire collection and production process for 100% traceability
  • Newborn bovine calf serum undergoes the same careful collection and processing procedures (including venipuncture) used for other bovine serum products
  • Sourced from the United States with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at or below -10°C
United States
Newborn Calf Bovine Serum
Sterile
Heat Inactivated
≤ 50 EU/mL
Yes
HyClone™
<10 days
Liquid
1000 mL
Sequential-filtered, 100 nm
1000 mL
Augmenting Cell Culture Growth
≤ 25 mg/dL
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers