missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Serum, New Zealand Origin
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
147.00 € - 176.46 €
Specifications
Age | <10 days |
---|---|
Country of Origin | New Zealand |
Product Type | Newborn Calf Bovine Serum |
Quantity | 1000 mL |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
16675775
|
Cytiva
SH30560.02HI |
Heat Inactivated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16685775
|
Cytiva
SH30560.02NI |
Non-Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16665775
|
Cytiva
SH30560.02 |
None |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases
Specifications
<10 days | |
Newborn Calf Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
≤ 25 mg/dL | |
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |
New Zealand | |
1000 mL | |
≤ 25 EU/mL | |
HyClone™ |