missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Penicllin Streptomycin Glutamine solution

Brand:  Cytiva SV30082.01


Product Code. 10319633

  • 23.90 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

-10° to -20° C
Broad Spectrum, Gram-Negative and Gram-Positive Bacteria
Liquid
100 mL
Pen/Strep/Glutamine Solution
1 X
0.1 μm Sterile-filtered
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HyClone Penicillin/Streptomycin/Glutamine Solution

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : streptomycin sulphate
  • UNKNOWN AQUATIC-Contains 3% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.