Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
82.80 € - 352.56 €
Specifications
Certifications/Compliance cGMP
View More Specs
Products 4
Product Code Brand Quantity With Additives Without Additives Product Type Price Quantity & Availability  
Product Code Brand Quantity With Additives Without Additives Product Type Price Quantity & Availability  
10494085
View Documents
Cytiva
SH30333.02
10 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS
294.00 €
10 L
11571911
View Documents
Cytiva
SH30359.LS
6 x 1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF CHO LS This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10443295
View Documents
Cytiva
SH30359.02
1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF-CHO LS
82.80 €
1000mL
10721694
View Documents
Cytiva
SH30333.01
5 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS
121.00 €
5 L
Description
Description
  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • Specifications
    Specifications
    cGMP
    Videos
    Safety and Handling
    Safety and Handling
    missing translation for 'shelfLife' : 36 Months