missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
Brand: Cytiva SH30258.01
17.84 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- QC-tested solution
Specifications
cGMP and ISO 9001:2000 Guidelines | |
10 X | |
Phosphate Buffered Saline (PBS) | |
No Calcium, No Magnesium, No Phenol Red | |
7.0 to 7.2 |
15° to 30°C | |
Liquid | |
0.1 μm Sterile-filtered | |
=1.0EU/mL | |
500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction