Learn More
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
Brand:  Cytiva SH30256.01
7.03 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 10462372
  • 19.65 € / 500mL
Edge
Explore more special offers
Description
Description
  • QC-tested solution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cGMP and ISO 9001:2000 Guidelines
1 X
Phosphate Buffered Saline (PBS)
No Calcium, No Magnesium, No Phenol Red
7.0 to 7.2
15° to 30°C
Liquid
0.1 μm Sterile-filtered
=1.0EU/mL
500 mL