missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ RPMI 1640 Media, Powder

Support cell culture growth and biomanufacturing processes. HyClone™ RPMI 1640 Media, Powder is manufactured using ISO 9001 certified processes.

Brand:  Cytiva SH30011.02

36.35 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 11958075

  • 42.20 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-quality classical media formulations in convenient packaging configurations
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Exacting manufacturing processes ensure lot-to-lot consistency
TRUSTED_SUSTAINABILITY
Specifications

Specifications

cGMP and ISO 9001:2000
Powder
Non-sterile
No Sodium Bicarbonate
2° to 8°C, away from light and moisture
RPMI 1640, with L-glutamine
Glutamine
2 x 5 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers