Learn More
Cytiva HyClone™ SFM4HEK293™ Media
36.40 € - 86.30 €
Specifications
Form Liquid
Certifications/Compliance cGMP
Description SFM4HEK293 Media
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10500283
View Documents
Cytiva
SH30521.02
1000 mL
86.30 €
1000mL
10061502
View Documents
Cytiva
SH30521.01
500 mL N/A N/A N/A
Description
Description
Support production of adenoviral vectors and recombinant proteins in HEK 239 cultures with HyClone™ SFM4HEK293™ Media, a versatile, protein-free cell culture media containing no animal-derived components.
Specifications
Specifications
Liquid
cGMP
SFM4HEK293 Media
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 12 Months
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cytiva HyClone™ SFM4HEK293™ Media