Learn More
Cytiva HyClone™ SFM4MAb Powdered Media
Increase process yields for manufacturing of human and humanized recombinant antibodies.
Brand: Cytiva SH30535.02
This item is not returnable.
View return policy
Description
SFM4MAb is a low protein formulation and is optimized for downstream purification using protein A, protein G, and other matrices to facilitate product recovery. The medium is formulated using proprietary lipid and phospholipid complexing process for enhanced stability and growth promotion of various cell types.
SFM4MAb is available in liquid and powder formats in user-friendly packaging.
- Designed for high cell yield and antibody production
- Complexed lipids for enhanced stability
- Allows for direct or sequential adaptation
- Optimized for protein A, protein G, and other purification matrices
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
2 to 8°C, away from light and moisture | |
Powder | |
Non-sterile | |
5 L |