missing translation for 'onlineSavingsMsg'
Learn More
HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva Product Code.: 11586271
In Stock
to see stock.
Add to basket

HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva

Product Code. 11586271
6 x 500 mL
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 500 mL
6 x 1 L
This item is not returnable. View return policy

Product Code. 11586271

Brand: HyClone products Cytiva SH30529.FS

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Eliminate endotoxin concerns with HyClone™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.

  • Endotoxin-free; contains no detectable gram-negative bacteria
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay
TRUSTED_SUSTAINABILITY

Specifications

pH 5.0 to 7.0
Description Water, Cell Culture Grade (Endotoxin-Free)
Endotoxin <0.005EU/mL
Quantity 6 x 500 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.