Learn More
HyClone™ Water, Molecular Biology Grade, Cytiva
Product Code. 10275262
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1 L
For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.
  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL