missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Water, Molecular Biology Grade, Cytiva

Product Code. 10787944
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.

  • Format: Liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 5.0 to 7.0
Quantity 500 mL
Chemical Name or Material Water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'