missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Water, Molecular Biology Grade, Cytiva

For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.

13.05 € - 165.00 €


Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10275262
View Documents
HyClone products Cytiva
SH30538.01
100 mL
13.05 €
100mL
In Stock
to see stock.
Edge
Add to basket
 
10787944
View Documents
HyClone products Cytiva
SH30538.02
500 mL
22.35 €
500mL
In Stock
to see stock.
Edge
Add to basket
 
10307052
View Documents
HyClone products Cytiva
SH30538.03
1000 mL
46.25 €
1000mL
In Stock
to see stock.
Edge
Add to basket
 
10247783
missing translation for 'viewDocuments'
HyClone products Cytiva
SH30538.FS
6 x 500 mL
132.00 €
Pack of 6
In Stock
to see stock.
Add to basket
 
10452915
missing translation for 'viewDocuments'
HyClone products Cytiva
SH30538.LS
6 x 1 L
165.00 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Format: Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

Recommended Storage : 2° to 30°C

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.