missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 27-602 Fully Nitrile Coated Heavy Duty Cotton Jersey Gloves

Product Code. 12670792
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Ansell's unique nitrile coating formulation offers resistance to sharp/abrasive materials, oil, and grease. The heavy duty design is durable, and certified clean for metals and motor manufacturing.

  • Ansell's unique nitrile formulation offers resistance to sharp and abrasive materials
  • Gloves provides a barrier against grease and oil that will not degrade
  • Guaranteed clean for metals and motor manufacturing
  • Three times more durable than heavy-duty leather
  • Stronger and more flexible than PVC
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Construction Cut and Sewn
Cuff Material Nitrile
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 9
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 27-602
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'