missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Buffer Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

174.00 € - 418.00 €

Specifications

Flash Point 11.00°C
Chemical Name or Material HYDRA-POINT™ Buffer Solution
Packaging Narrow Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15547634
View Documents
Avantor J.T.Baker
6282-01
500 mL Narrow Mouth Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15557634
View Documents
Avantor J.T.Baker
6282-02
1 L Narrow Mouth Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

    For Karl Fischer titration
Specifications

Specifications

11.00°C
HYDRA-POINT™ Buffer Solution
Narrow Mouth Amber Glass Bottle
BAKER ANALYZED™ Reagent
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.