missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Buffer Base, Honeywell Fluka™
Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid)
106.00 € - 553.23 €
Specifications
Flash Point | 11°C (51.8°F) |
---|---|
Chemical Name or Material | HYDRANAL™-Buffer Base |
CAS | 64-17-5, 67-56-1, 69-72-7, 121-44-8 |
MDL Number | MFCD00147178 |
UN Number | UN1993BE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15695160
|
Honeywell-Hydranal
37859-1L |
1 L |
106.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15615170
|
Honeywell-Hydranal
37859-6X1L |
6 x 1 L |
553.23 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
11°C (51.8°F) | |
64-17-5, 67-56-1, 69-72-7, 121-44-8 | |
UN1993BE |
HYDRANAL™-Buffer Base | |
MFCD00147178 |
Safety and Handling
P210-P280-P304 + P340 + P312-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol, salicylic acid, triethylamine