Learn More
HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Brand:  Honeywell Fluka 34806-6X1L
820.28 EUR valid until 2025-12-31
Use promo code "24351" to get your promotional price.
 View more versions of this product
Product Code. 15644450
  • 863.45 € / Pack of 6
Explore more special offers
Specifications
HYDRANAL™-Composite 2
90°C (194°F)
111-90-0, 288-32-4, 7446-09-5, 7553-56-2, 693-98-1, 68007-08-9
1.11g/mL
6 x 1 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • HYDRANAL-Composite 2
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : imidazole, sulphur dioxide, iodine, 2-Methylimidazole, 1H-Imidazole monohydriodide