missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Composite 5, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Brand: Honeywell Fluka 34805-4X2.5L
1222.14 EUR valid until 2025-12-31
Use promo code "24351" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
HYDRANAL™-Composite 5 | |
90°C (194°F) | |
111-90-0, 7553-56-2, 288-32-4, 7446-09-5, 68007-08-9, 693-98-1 |
1.15g/mL | |
4 x 2.5 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Composite 5
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sulphur dioxide, 1H-Imidazole monohydriodide, iodine, imidazole, 2-Methylimidazole
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction