missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™
Brand: Honeywell-Hydranal 34816-2.5L
407.10 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
HYDRANAL™-Composite 5 K | |
90°C (194°F) | |
111-90-0, 7553-56-2, 288-32-4, 693-98-1, 7446-09-5, 68007-08-9 |
1.15g/mL | |
2.5 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL Composite 5 K
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : iodine, imidazole, sulphur dioxide, 1H-Imidazole monohydriodide, 2-Methylimidazole
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction