missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™ - Coulomat AF 7 |
| Name Note | For coulometer AF7 |
| Boiling Point | 63°C |
| CAS Min % | ≥1.0000%, ≥5.0000% |
| CAS Max % | <10.0000%, <5.0000% |
| CAS | 288-32-4, 68007-08-9, 7446-09-5 |
| Density | 0.881 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | 11°C |
| Packaging | Glass Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction