Learn More
HYDRANAL™ - Coulomat AG-H, Reagent for coulometric KF titration in long-chained hydrocarbons (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™
149.68 € - 778.81 €
Specifications
UN Number UN1992
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15604510
View Documents
Honeywell-Hydranal
34843-500ML
500 mL
149.68 €
500mL
15614510
View Documents
Honeywell Fluka
34843-6X500ML
6 x 500 mL
778.81 €
Pack of 6
Specifications
UN1992
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
HYDRANAL™ - Coulomat AG-H, Reagent for coulometric KF titration in long-chained hydrocarbons (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™