missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™

Brand:  Honeywell Fluka 34836-6X500ML

626.08 EUR valid until 2024-12-31
Use promo code "21961" to get your promotional price.



 View more versions of this product

Product Code. 15644500

  • 698.57 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Coulomat AG
6 x 500 mL
UN1230
14°C (57.2°F)
67-56-1, 111-42-2, 7446-09-5, 288-32-4, 68007-08-9
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • hydranal coulomat AG

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, 2,2'-iminodiethanol; diethanolamine, imidazole, sulphur dioxide, 1H-imidazole monohydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers