missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™
Brand: Honeywell-Hydranal 34836-1L
224.39 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
Specifications
HYDRANAL™-Coulomat AG | |
1 L | |
UN1230 |
14°C (57.2°F) | |
67-56-1, 111-42-2, 7446-09-5, 288-32-4, 68007-08-9 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, 2,2'-iminodiethanol; diethanolamine, imidazole, sulphur dioxide, 1H-Imidazole monohydriodi