missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat AG-Oven, Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm, Honeywell Fluka
Brand: Honeywell-Hydranal 34739-6X500ML
758.75 EUR valid until 2025-12-31
Use promo code "24351" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
Specifications
HYDRANAL™ - Coulomat AG-Oven | |
0.980 g/cm3 | |
6 x 500 mL | |
<70.0000%, <30.0000%, <20.0000%, <10.0000%, <10.0000%, <10.0000% | |
64°C | |
UN1230 |
Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm | |
13°C | |
≥50.0000%, ≥20.0000%, ≥10.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000% | |
57-55-6, 111-42-2, 288-32-4, 7446-09-5, 68007-08-9 | |
Glass Bottle | |
Karl Fischer |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Coulomat AG-Oven
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol 2,2'-iminodiethanol; diethanolamine imidazole sulphur dioxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction