missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat CG-K, Reagent for coulometric KF titration in ketones (catholyte solution), Honeywell Fluka™

292.94 € - 1546.27 €

Specifications

CAS 123-39-7, 288-32-4, 7446-09-5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15604490
View Documents
Honeywell-Hydranal
34821-50ML
10 x 5 mL Ampules
292.94 €
50mL
Please to purchase this item. Need a web account? Register with us today!
15614490
View Documents
Honeywell-Hydranal
34821-6X50ML
1 Kit
1546.27 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

123-39-7, 288-32-4, 7446-09-5
Videos
Safety and Handling

Safety and Handling

P201-P302 + P352 + P312-P305 + P351 + P338-P308 + P313-P337 + P313

Product Identifier
  • HYDRANAL-Coulomat CG-K

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : N-methylformamide, tetrahydro-2-furylmethanol; tetrahydrofurfuryl alcohol, imidazole, sulphur dioxid

SDS
Documents

Documents

Product Certifications