missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm
Brand: Honeywell-Hydranal 34868-100ML
43.39 EUR valid until 2024-12-31
Use promo code "21961" to get your promotional price.
This item is not returnable.
View return policy
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
HYDRANAL™-Coulomat Oil | |
100 mL | |
UN1992 |
8°C (46.4°F) | |
68007-08-9, 67-66-3, 95-47-6 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Coulomat Oil
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, o-xylene,trichloromethane; chloroform, imidazole,1H-Imidazole monohydriodide, sulphur di