missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™

Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm

Brand:  Honeywell Fluka 34868-6X500ML


 View more versions of this product

Product Code. 15694620

  • 731.99 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Coulomat Oil
8°C (46.4°F)
6 x 500 mL
UN1992
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL-Coulomat Oil

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, o-xylene,trichloromethane; chloroform, imidazole,1H-Imidazole monohydriodide, sulphur di

SDS
Documents

Documents

Certificates
Special Offers

Special Offers