missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Humidity absorber, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
6 x 1 kg
Unit Size:
1kg
500g
Pack of 6
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Humidity absorber |
| CAS | 7631-86-9 |
| Quantity | 6 x 1 kg |
| UN Number | NONH for all modes of transport |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction