missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Humidity absorber, Honeywell Fluka™

Product Code. p-7072129 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
6 x 1 kg
Unit Size:
1kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15634410

Brand: Honeywell-Hydranal 34788500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Drying agent for air and gases for Karl Fischer titration (with indicator)

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Humidity absorber
CAS 7631-86-9
Quantity 500 g
UN Number NONH for all modes of transport
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.