missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Methanol Rapid, Reagent for accelerated volumetric one-component KF titration (working medium), Honeywell Fluka™

Medium for accelerated volumetric one-component Karl Fischer titration.

63.53 € - 519.18 €

Specifications

UN Number UN1230
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15605160
View Documents
Honeywell-Hydranal
37817-1L
1 L
63.53 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15615160
View Documents
Honeywell-Hydranal
37817-2.5L
2.5 L
149.76 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15635160
View Documents
Honeywell Fluka
37817-6X1L
6 x 1 L
322.30 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15625160
View Documents
Honeywell Fluka
37817-4X2.5L
4 x 2.5 L
519.18 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

UN1230
Videos
Safety and Handling

Safety and Handling

P201-P210-P280-P308 + P311-P370 + P378-P403 + P235

Product Identifier
  • HYDRANAL-Methanol Rapid

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, sulphur dioxide, imidazole

SDS
Documents

Documents

Product Certifications