missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Coulomat AG-FI, Honeywell Fluka™

Anolyte for coulometric Karl Fischer titrations, for cells with and without diaphragm

Brand:  Honeywell Fluka 34433-500ML


 View more versions of this product

Product Code. 16822370

  • 158.94 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Methanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - NEXTGEN Coulomat AG-FI
0.893 g/cm3 (at 20°C)
500 mL
≥60%
Glass Bottle
Liquid
UN1230
Yellow
14°C
67-56-1
64°C to 65°C (Methanol)
-98°C (Methanol)
≤90%
Completely Miscible in Water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL NEXTGEN Coulomat AG-FI

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, 2-ethylimidazole, 2-butylaminoethanol, sulphur dioxide, 2-ethyl imidazole hydroiodide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.