missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - NEXTGEN Coulomat C-FA, Honeywell Fluka™
Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances).
415.68 € - 2194.15 €
Specifications
Density | 0.906 g/cm3 |
---|---|
Flash Point | 12°C to 13°C |
Chemical Name or Material | HYDRANAL™ - NEXTGEN Coulomat C-FA |
Color | Orange |
CAS | 75-05-8, Confidential Stabilizer 2, 75-75-2, Confidential Stabilizer 1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
17584946
|
Honeywell Fluka
34470-50ML |
50 mL | Ampoule |
415.68 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17594946
|
Honeywell Fluka
34470-6X50ML |
6 x 50 mL | Ampoule |
2194.15 €
6 bottles |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
- Comprehensive product portfolio with 60 available products
- 40 years of innovation – check our NEXTGEN reagents HERE
- Dedicated technical service and support
- More than 800 Application Notes available on our website
Specifications
0.906 g/cm3 | |
HYDRANAL™ - NEXTGEN Coulomat C-FA | |
75-05-8, Confidential Stabilizer 2, 75-75-2, Confidential Stabilizer 1 | |
≥10.00, ≥60.00, ≥2.00, ≥2.00 | |
Liquid | |
Soluble in water |
12°C to 13°C | |
Orange | |
5.5 | |
Ampoule | |
UN2924 |
Safety and Handling
missing translation for 'shelfLife' : 720 days
Recommended Storage : Room Temperature
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL NEXTGEN Coulomat C-FA
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile, methanesulphonic acid, 1-Ethylimidazole