Learn More
HYDRANAL™-Sodium tartrate dihydrate, Honeywell Fluka™
Test substance for volumetric Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)
Brand:  Honeywell-Hydranal 34803-6X100G
 View more versions of this product
Product Code. 15664430
  • 335.54 € / Pack of 6
Explore more special offers
Description
Description
Advantages of HYDRANAL- Auxiliaries
  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HYDRANAL™-Sodium tartrate dihydrate
6 x 100 g
6106-24-7
NONH for all modes of transport